Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008792.4 | NP_032818.1 | 4 | 848 | ||
GenBank mRNA | AF483512.1 | - | 4 | 490 | ||
AF483513.1 | - | 4 | 490 | |||
AK133620.1 | - | 4 | 849 | |||
AK158493.1 | - | 4 | 796 | |||
AK158598.1 | - | 4 | 910 | |||
AK161585.1 | - | 4 | 789 | |||
BC052013.1 | - | 4 | 765 | 5707923 | ||
BC057348.1 | - | 4 | 803 | 6827695 | ||
M55669.1 | - | 4 | 574 |