Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281934.1 | NP_001268863.1 | 6 | 1701 | ||
NM_008794.2 | NP_032820.2 | 6 | 1437 | |||
GenBank mRNA | AK148416.1 | - | 6 | 1022 | ||
AK151980.1 | - | 6 | 1024 | |||
AK165389.1 | - | 6 | 1043 | |||
BC006730.1 | - | 5 | 938 | 3962441 | ||
BC007322.1 | - | 6 | 1300 | |||
BC056456.1 | - | 6 | 1437 | |||
U48830.1 | - | 6 | 1009 |