Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008795.2 | NP_032821.1 | 7, 8 | 908 | ||
GenBank mRNA | AK004998.1 | - | 7, 8 | 852 | ||
AK171569.1 | - | 7, 8 | 689 | |||
BC119617.1 | - | 8, 9 | 784 | 40061728 | ||
BC119618.1 | - | 2, 3 | 175 | |||
X69026.1 | - | 7, 8 | 925 |