Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206567.1 | NP_001193496.1 | 9 | 2114 | ||
NM_005531.2 | NP_005522.2 | 9 | 2114 | |||
XM_005245127.3 | XP_005245184.1 | 11 | 2599 | |||
XM_006711290.1 | XP_006711353.1 | 8 | 1946 | |||
XM_017001149.1 | XP_016856638.1 | 9 | 2263 | |||
GenBank mRNA | AB208989.1 | - | 9 | 2084 | ||
AF208043.1 | - | 9 | 2088 | |||
AK094968.1 | - | 7 | 1487 | |||
AK296228.1 | - | 9 | 1997 | |||
AK307957.1 | - | 9 | 1895 | |||
AY138863.1 | - | 8 | 1824 | |||
BC017059.1 | - | 9 | 2081 | 3914632 | ||
M63838.1 | - | 9 | 2088 |