Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011051.3 | NP_035181.1 | 6 | 651 | ||
GenBank mRNA | AK008610.1 | - | 6 | 575 | ||
AK028223.1 | - | 6 | 606 | |||
AK078044.1 | - | 6 | 579 | |||
AK150335.1 | - | 6 | 569 | |||
BC040079.1 | - | 6 | 602 | 3156800 | ||
U49112.1 | - | 6 | 588 | |||
X14938.1 | - | 5 | 418 |