Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011053.2 | NP_035183.2 | 11 | 1452 | ||
GenBank mRNA | AF055668.1 | - | 2 | 155 | ||
AK129080.1 | - | 11 | 1450 | |||
AK141450.1 | - | 11 | 1453 | |||
AK147950.1 | - | 11 | 1437 | |||
AK161803.1 | - | 11 | 1442 | |||
BC010844.1 | - | 11 | 1432 | |||
BC070468.1 | - | 11 | 1439 | 30628812 |