Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130080.2 | NP_001123552.1 | 3 | 235 | ||
NM_001288952.1 | NP_001275881.1 | 4 | 288 | |||
NM_001288954.1 | NP_001275883.1 | 4 | 288 | |||
NM_001288956.1 | NP_001275885.1 | 3 | 201 | |||
NM_001288957.1 | NP_001275886.1 | 3 | 300 | |||
NM_001288958.1 | NP_001275887.1 | 3 | 165 | |||
NM_001288959.1 | NP_001275888.1 | 2 | 86 | |||
NM_001288960.1 | NP_001275889.1 | 2 | 86 | |||
NM_001288995.1 | NP_001275924.1 | 3 | 201 | |||
NM_005532.4 | NP_005523.3 | 3 | 235 | |||
GenBank mRNA | AJ294851.2 | - | 3 | 196 | ||
AK095039.1 | - | 1 | 2022 | |||
AK289535.1 | - | 3 | 233 | |||
AK312089.1 | - | 3 | 232 | |||
AY126456.1 | - | 3 | 159 | |||
AY126457.1 | - | 2 | 80 | |||
AY126458.1 | - | 2 | 80 | |||
BC015492.1 | - | 3 | 300 | 3915505 | ||
BF971643.1 | - | 4 | 267 | |||
BG483267.1 | - | 3 | 199 | |||
BM756566.1 | - | 2 | 83 | |||
BM841506.1 | - | 3 | 198 | |||
BM841596.1 | - | 3 | 232 | |||
BP317624.1 | - | 4 | 285 | |||
BT006781.1 | - | 2 | 112 | |||
BU601451.1 | - | 3 | 235 | |||
BU928223.1 | - | 2 | 87 | |||
BX647330.1 | - | 3 | 232 | |||
CB993374.1 | - | 2 | 110 | |||
X67325.1 | - | 2 | 166 |