Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024959.2 | NP_001020130.1 | 6 | 1526 | ||
NM_001024960.2 | NP_001020131.1 | 6 | 1381 | |||
NM_001198780.1 | NP_001185709.1 | 6 | 1380 | |||
NM_005718.4 | NP_005709.1 | 6 | 1892 | |||
GenBank mRNA | AK074205.1 | - | 1 | 1995 | ||
BC012596.2 | - | 5 | 1311 |