Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163748.1 | NP_001157220.1 | 4 | 286 | ||
NM_008804.4 | NP_032830.3 | 4 | 286 | |||
GenBank mRNA | AF031147.1 | - | 4 | 241 | ||
AF068247.1 | - | 4 | 250 | |||
AK030509.1 | - | 4 | 270 | |||
AK168143.1 | - | 4 | 268 | |||
BC025882.1 | - | 4 | 244 | |||
BC038675.1 | - | 4 | 235 | |||
BC046915.1 | - | 4 | 235 | |||
BC061163.1 | - | 4 | 266 | 6533665 | ||
BY283127.1 | - | 4 | 287 |