Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF190928.1 | - | 1 | 111 | ||
AK080735.1 | - | 2 | 257 | |||
BC014723.1 | - | 2 | 226 | |||
BC082288.1 | - | 2 | 214 | 6490841 |