Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083316.1 | NP_001076785.1 | 20 | 2908 | ||
NM_011058.2 | NP_035188.2 | 20 | 2891 | |||
GenBank mRNA | AK136490.1 | - | 20 | 2877 | ||
AK147267.1 | - | 20 | 2840 | |||
AK163952.1 | - | 20 | 2955 | |||
BC053036.1 | - | 20 | 2908 | 5704645 | ||
M57683.1 | - | 20 | 2790 | |||
M84607.1 | - | 20 | 2901 |