Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083316.1 | NP_001076785.1 | 13 | 2054 | ||
NM_011058.2 | NP_035188.2 | 13 | 2037 | |||
GenBank mRNA | AK035501.1 | - | 13 | 1995 | ||
AK081664.1 | - | 13 | 2036 | |||
AK136490.1 | - | 13 | 2023 | |||
AK145097.1 | - | 13 | 2638 | |||
AK147267.1 | - | 13 | 1986 | |||
AK163952.1 | - | 13 | 2101 | |||
BC053036.1 | - | 13 | 2054 | 5704645 | ||
M57683.1 | - | 13 | 1936 | |||
M84607.1 | - | 13 | 2047 |