Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008812.2 | NP_032838.2 | 2 | 252 | ||
GenBank mRNA | AK148156.1 | - | 2 | 239 | ||
AK154814.1 | - | 2 | 238 | |||
AK158382.1 | - | 2 | 239 | |||
AK171275.1 | - | 2 | 239 | |||
BB850871.1 | - | 2 | 254 | |||
BC040350.1 | - | 2 | 194 | 4192742 | ||
BC049947.1 | - | 2 | 225 | 3600978 | ||
D16580.1 | - | 2 | 185 |