Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136077.2 | NP_001129549.1 | 18 | 1820 | ||
NM_001285994.1 | NP_001272923.1 | 17 | 1664 | |||
NM_001285995.1 | NP_001272924.1 | 18 | 1820 | |||
NM_015744.3 | NP_056559.2 | 17 | 1664 | |||
GenBank mRNA | AF123542.1 | - | 17 | 1520 | ||
AK088491.1 | - | 18 | 1692 | |||
AK161144.1 | - | 17 | 1537 | |||
AK163881.1 | - | 17 | 1538 | |||
BC003264.1 | - | 17 | 1541 | 3499038 | ||
BC058759.1 | - | 17 | 1627 | 6491897 | ||
EU131009.1 | - | 18 | 1635 | |||
EU131010.1 | - | 17 | 1479 |