Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286502.1 | NP_001273431.1 | 4 | 440 | ||
NM_018863.4 | NP_061351.2 | 4 | 631 | |||
GenBank mRNA | AF026537.1 | - | 4 | 526 | ||
AF483510.1 | - | 2 | 323 | |||
AF483511.1 | - | 2 | 323 | |||
AK133597.1 | - | 4 | 632 | |||
BC137571.1 | - | 4 | 470 | 8860590 | ||
BC137572.1 | - | 4 | 470 | 8860591 |