Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Per3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289877.1 | NP_001276806.1 | 2 | 651 | ||
NM_001289878.1 | NP_001276807.1 | 2 | 651 | |||
NM_011067.3 | NP_035197.2 | 2 | 651 | |||
GenBank mRNA | AF050182.1 | - | 2 | 575 | ||
AK036345.1 | - | 2 | 568 | |||
AK081361.1 | - | 1 | 177 | |||
BC141032.1 | - | 2 | 249 | 9055644 | ||
BC145178.1 | - | 2 | 249 |