Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008823.3 | NP_032849.2 | 4 | 534 | ||
GenBank mRNA | AK150212.1 | - | 4 | 495 | ||
AK152782.1 | - | 4 | 485 | |||
AK154306.1 | - | 4 | 518 | |||
AK159291.1 | - | 4 | 535 | |||
AK159609.1 | - | 4 | 485 | |||
AK159853.1 | - | 4 | 485 | |||
AK171374.1 | - | 4 | 518 | |||
BC132369.1 | - | 4 | 518 | 40130646 | ||
BC138306.1 | - | 4 | 518 | 8861326 | ||
X12905.1 | - | 3 | 383 |