Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031683.3 | NP_001026853.1 | 2 | 1092 | ||
NM_001289758.1 | NP_001276687.1 | 3 | 1321 | |||
NM_001289759.1 | NP_001276688.1 | 3 | 1214 | |||
NM_001549.5 | NP_001540.2 | 2 | 1203 | |||
GenBank mRNA | AF026939.1 | - | 2 | 1042 | ||
AF083470.1 | - | 2 | 1026 | |||
AK290427.1 | - | 2 | 1113 | |||
AK297137.1 | - | 3 | 1012 | |||
AK312675.1 | - | 2 | 1023 | |||
BC001383.2 | - | 2 | 1025 | 3050764 | ||
BC004977.1 | - | 2 | 998 | 2906188 | ||
BT007284.1 | - | 1 | 948 | |||
U52513.1 | - | 2 | 1009 |