Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008828.3 | NP_032854.2 | 3 | 375 | ||
GenBank mRNA | AK133877.1 | - | 3 | 297 | ||
AK145846.1 | - | 3 | 311 | |||
AK164440.1 | - | 3 | 297 | |||
AK167441.1 | - | 3 | 269 | |||
AK167459.1 | - | 3 | 269 | |||
AK167710.1 | - | 3 | 268 | |||
AK180821.1 | - | 1 | 29 | |||
AK206980.1 | - | 2 | 147 | |||
BC023819.1 | - | 3 | 282 | |||
BC032167.1 | - | 3 | 282 | |||
BC083355.1 | - | 3 | 267 | 6828087 | ||
BC108372.1 | - | 3 | 279 | 5060103 | ||
M15668.1 | - | 3 | 246 |