Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008828.3 | NP_032854.2 | 2 | 255 | ||
GenBank mRNA | AK133877.1 | - | 2 | 177 | ||
AK145846.1 | - | 2 | 191 | |||
AK164440.1 | - | 2 | 177 | |||
AK167441.1 | - | 2 | 149 | |||
AK167459.1 | - | 2 | 149 | |||
AK167710.1 | - | 2 | 148 | |||
AK211934.1 | - | 1 | 27 | |||
BC023819.1 | - | 2 | 162 | |||
BC032167.1 | - | 2 | 162 | |||
BC083355.1 | - | 2 | 147 | 6828087 | ||
BC108372.1 | - | 2 | 159 | 5060103 | ||
BY292706.1 | - | 2 | 256 | |||
EF437325.1 | - | 2 | 119 | |||
M15668.1 | - | 2 | 126 |