Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008828.3 | NP_032854.2 | 6 | 715 | ||
GenBank mRNA | AK133877.1 | - | 6 | 637 | ||
AK145846.1 | - | 6 | 651 | |||
AK164440.1 | - | 6 | 637 | |||
AK167441.1 | - | 6 | 609 | |||
AK167459.1 | - | 6 | 609 | |||
AK167710.1 | - | 6 | 608 | |||
AK207010.1 | - | 2 | 134 | |||
BC023819.1 | - | 6 | 621 | |||
BC032167.1 | - | 6 | 621 | |||
BC083355.1 | - | 6 | 607 | 6828087 | ||
BC108372.1 | - | 6 | 619 | 5060103 | ||
M15668.1 | - | 6 | 586 |