Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031683.3 | NP_001026853.1 | 2 | 755 | ||
NM_001289758.1 | NP_001276687.1 | 3 | 984 | |||
NM_001289759.1 | NP_001276688.1 | 3 | 877 | |||
NM_001549.5 | NP_001540.2 | 2 | 866 | |||
GenBank mRNA | AF026939.1 | - | 2 | 705 | ||
AF083470.1 | - | 2 | 689 | |||
AK290427.1 | - | 2 | 776 | |||
AK297137.1 | - | 3 | 675 | |||
AK312675.1 | - | 2 | 686 | |||
BC001383.2 | - | 2 | 688 | 3050764 | ||
BC004977.1 | - | 2 | 661 | 2906188 | ||
BT007284.1 | - | 1 | 611 | |||
BU509201.1 | - | 3 | 817 | |||
U52513.1 | - | 2 | 672 |