Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011075.2 | NP_035205.1 | 15 | 1917 | ||
GenBank mRNA | AK030328.1 | - | 14 | 1860 | ||
BC141363.1 | - | 15 | 1874 | 9055975 | ||
BC150811.1 | - | 15 | 1874 | 9087722 | ||
M14757.1 | - | 15 | 1877 |