Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008831.4 | NP_032857.1 | 3 | 338 | ||
GenBank mRNA | AK002714.1 | - | 2 | 284 | ||
AK010619.1 | - | 3 | 281 | |||
AK150956.1 | - | 3 | 283 | |||
AK151073.1 | - | 3 | 283 | |||
AK152933.1 | - | 3 | 285 | |||
AK160490.1 | - | 3 | 277 | |||
AK168656.1 | - | 3 | 277 | |||
BC083354.1 | - | 2 | 260 | 5715690 | ||
BC089034.1 | - | 2 | 262 | 30675988 | ||
X78682.1 | - | 2 | 266 |