Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163467.1 | NP_001156939.1 | 5 | 402 | ||
NM_001163469.1 | NP_001156941.1 | 5 | 808 | |||
NM_013629.2 | NP_038657.2 | 5 | 808 | |||
GenBank mRNA | AJ133721.1 | - | 5 | 402 | ||
AJ242864.1 | - | 5 | 808 | |||
AK019146.1 | - | 5 | 712 | |||
AK131629.1 | - | 699 | ||||
BC021147.1 | - | 5 | 680 | |||
BC050825.1 | - | 5 | 620 | 6518647 | ||
BC057340.1 | - | 5 | 694 | 6827816 | ||
BC060177.1 | - | 5 | 694 | 6827816 |