Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000605.3 | NP_000596.2 | 1 | 399 | ||
GenBank mRNA | AY255838.1 | - | 1 | 265 | ||
BC074936.2 | - | 1 | 375 | 30915269 | ||
BC074937.2 | - | 1 | 375 | 30915492 | ||
BC104163.1 | - | 1 | 376 | 40029595 | ||
BC104164.1 | - | 1 | 376 | 40029596 | ||
CR541921.1 | - | 1 | 331 | |||
JN591568.1 | - | 1 | 331 | |||
JN591569.1 | - | 1 | 331 | |||
JN591570.1 | - | 1 | 331 | |||
JN848522.1 | - | 1 | 331 | |||
M54886.1 | - | 1 | 391 | |||
V00544.1 | - | 1 | 391 | |||
V00548.1 | - | 1 | 313 | |||
V00549.1 | - | 1 | 391 |