Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008838.1 | NP_032864.1 | 4, 5 | 564 | ||
GenBank mRNA | AK132796.1 | - | 4, 5 | 570 | ||
AK171083.1 | - | 4, 5 | 552 | |||
BC028862.1 | - | 4, 5 | 553 | 3673399 | ||
D50264.1 | - | 4, 5 | 564 |