Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011083.2 | NP_035213.2 | 9 | 1937 | ||
GenBank mRNA | AK140908.1 | - | 10 | 2017 | ||
AK142284.1 | - | 10 | 1966 | |||
BC096553.1 | - | 8 | 1371 | |||
U52193.1 | - | 10 | 1843 | |||
U55772.1 | - | 8 | 1253 |