Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011083.2 | NP_035213.2 | 19 | 3416 | ||
GenBank mRNA | AK138724.1 | - | 5 | 690 | ||
BC096553.1 | - | 18 | 2851 | |||
U52193.1 | - | 20 | 3322 | |||
U55772.1 | - | 18 | 2732 |