Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011084.2 | NP_035214.2 | 10 | 1475 | ||
NM_207683.2 | NP_997566.1 | 4221 | ||||
GenBank mRNA | AB008791.1 | - | 4221 | |||
AB008792.1 | - | 10 | 1470 | |||
AK054271.1 | - | 10 | 1351 | |||
BC150813.1 | - | 4213 | 9087724 |