Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029837.2 | NP_001025008.2 | 8 | 1275 | ||
NM_001164049.1 | NP_001157521.1 | 8 | 1302 | |||
NM_001164050.1 | NP_001157522.1 | 8 | 1302 | |||
NM_001164051.1 | NP_001157523.1 | 8 | 1326 | |||
NM_001164052.1 | NP_001157524.1 | 6 | 1029 | |||
NM_008840.3 | NP_032866.2 | 9 | 1477 | |||
GenBank mRNA | AK040867.1 | - | 8 | 1237 | ||
AK149935.1 | - | 9 | 1376 | |||
AK150649.1 | - | 8 | 1203 | |||
AK151482.1 | - | 8 | 1203 | |||
AK152661.1 | - | 8 | 1203 | |||
AK153304.1 | - | 8 | 1237 | |||
AK155835.1 | - | 8 | 1188 | |||
AK171025.1 | - | 8 | 1285 | |||
AK172222.1 | - | 8 | 1179 | |||
BC035203.1 | - | 8 | 1292 | 4192906 | ||
CT010349.1 | - | 6 | 997 | |||
U86587.1 | - | 6 | 997 |