Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029837.2 | NP_001025008.2 | 21, 22 | 3013 | ||
NM_001164049.1 | NP_001157521.1 | 21, 22 | 3040 | |||
NM_001164050.1 | NP_001157522.1 | 21, 22 | 3034 | |||
NM_001164051.1 | NP_001157523.1 | 21, 22 | 3061 | |||
NM_001164052.1 | NP_001157524.1 | 19, 20 | 2755 | |||
NM_008840.3 | NP_032866.2 | 22, 23 | 3206 | |||
GenBank mRNA | AK040867.1 | - | 21, 22 | 2975 | ||
AK149935.1 | - | 22, 23 | 3105 | |||
AK153304.1 | - | 21, 22 | 2954 | |||
AK171025.1 | - | 21, 22 | 3020 | |||
AK172222.1 | - | 21, 22 | 2917 | |||
BC035203.1 | - | 21, 22 | 3024 | 4192906 | ||
CT010349.1 | - | 19, 20 | 2729 | |||
U86587.1 | - | 19, 20 | 2723 |