Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024955.2 | NP_001020126.1 | 4 | 812 | ||
NM_001077495.2 | NP_001070963.1 | 10 | 1901 | |||
GenBank mRNA | AK046259.1 | - | 4 | 814 | ||
BC026146.1 | - | 10 | 1882 | 3979333 | ||
BC051106.1 | - | 4 | 512 | 5031261 | ||
CB519962.1 | - | 4 | 444 | |||
M60651.1 | - | 10 | 1836 | |||
U50413.1 | - | 10 | 1854 |