Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008841.2 | NP_032867.2 | 9 | 1520 | ||
GenBank mRNA | AK146195.1 | - | 9 | 1510 | ||
AK148122.1 | - | 9 | 1517 | |||
AK219027.1 | - | 1 | 25 | |||
BC006796.1 | - | 9 | 1511 | 2581969 | ||
BC085501.1 | - | 8 | 1443 | 6816009 | ||
Y13569.1 | - | 9 | 1489 |