Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5k1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008846.2 | NP_032872.1 | 8 | 962 | ||
GenBank mRNA | AF048695.1 | - | 6 | 811 | ||
AK133809.1 | - | 8 | 957 | |||
AK146100.1 | - | 8 | 962 | |||
AK149354.1 | - | 8 | 962 | |||
AK164552.1 | - | 8 | 945 | |||
BC034864.1 | - | 8 | 976 | 3326897 | ||
D86176.1 | - | 7 | 895 |