Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5k1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293707.1 | NP_001280636.1 | 8 | 1416 | ||
NM_008847.3 | NP_032873.2 | 8 | 1413 | |||
GenBank mRNA | AK098097.1 | - | 8 | 1370 | ||
AK150898.1 | - | 8 | 1373 | |||
AK151984.1 | - | 8 | 1373 | |||
AK158062.1 | - | 8 | 1358 | |||
BC031774.1 | - | 8 | 1341 | 4503697 | ||
D86177.1 | - | 8 | 1203 |