Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pira1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011087.1 | NP_035217.1 | 2648 | |||
GenBank mRNA | U96682.1 | - | 2648 |
Gene Symbol: Gm10693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137448.1 | - | 7 | 3251 |
Gene Symbol: Pira2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011089.2 | NP_035219.2 | 10 | 2667 | ||
GenBank mRNA | AK140776.1 | - | 10 | 2673 | ||
AK172046.1 | - | 2668 |
Gene Symbol: Pira6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289428.1 | NP_001276357.1 | 2647 |