Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pira1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011087.1 | NP_035217.1 | 2494 | |||
GenBank mRNA | U96682.1 | - | 2494 |
Gene Symbol: Pira2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011089.2 | NP_035219.2 | 10 | 2513 | ||
GenBank mRNA | AK140776.1 | - | 10 | 2519 | ||
AK172046.1 | - | 2514 |