Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008852.4 | NP_032878.1 | 3 | 390 | ||
GenBank mRNA | AB221559.1 | - | 2 | 258 | ||
AF005772.1 | - | 3 | 386 | |||
BC120844.1 | - | 2 | 279 | 40129767 | ||
BC137810.1 | - | 2 | 279 | 8860829 |