Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253883.1 | NP_001240812.1 | 6 | 1100 | ||
NM_011099.3 | NP_035229.2 | 6 | 1100 | |||
GenBank mRNA | AK083076.1 | - | 6 | 893 | ||
AK135397.1 | - | 6 | 893 | |||
AK151724.1 | - | 6 | 966 | |||
AK153483.1 | - | 6 | 966 | |||
AK154695.1 | - | 6 | 893 | |||
AK155110.1 | - | 6 | 893 | |||
AK155655.1 | - | 6 | 896 | |||
AK168943.1 | - | 6 | 893 | |||
AK169872.1 | - | 6 | 872 | |||
AK170892.1 | - | 6 | 896 | |||
AK171023.1 | - | 6 | 893 | |||
AK171033.1 | - | 6 | 896 | |||
AK171106.1 | - | 6 | 897 | |||
AK211250.1 | - | 1 | 56 | |||
BC016619.1 | - | 6 | 860 | 3598842 | ||
BC094663.1 | - | 6 | 839 | 6829532 | ||
D38379.1 | - | 6 | 834 | |||
X97047.1 | - | 5 | 811 |