Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021057.2 | NP_066401.2 | 1 | 494 | ||
GenBank mRNA | BC074991.2 | - | 1 | 492 | 30915521 | |
BC074992.2 | - | 1 | 492 | 30915307 | ||
BC113994.1 | - | 1 | 494 | 40084821 | ||
BC114000.1 | - | 1 | 494 | 40084820 |
Gene Symbol: IFNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002171.2 | NP_002162.1 | 1 | 500 | ||
GenBank mRNA | BC069409.1 | - | 1 | 500 | 7262186 | |
BC103972.2 | - | 1 | 500 | 40015081 | ||
BC103973.1 | - | 1 | 500 | 40015082 | ||
V00551.1 | - | 1 | 500 |