Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277898.1 | NP_001264827.1 | 4 | 479 | ||
NM_008854.5 | NP_032880.1 | 4 | 522 | |||
GenBank mRNA | AF239743.1 | - | 4 | 479 | ||
BC003238.1 | - | 4 | 512 | 3497908 | ||
BC054834.1 | - | 4 | 507 | 6439034 | ||
BU059097.1 | - | 4 | 472 | |||
JN566056.1 | - | 4 | 436 | |||
M12303.1 | - | 4 | 499 |