Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164198.1 | NP_001157670.1 | 9 | 1125 | ||
NM_001164199.1 | NP_001157671.1 | 9 | 998 | |||
NM_001164200.1 | NP_001157672.1 | 9 | 1090 | |||
NM_011100.4 | NP_035230.1 | 9 | 1129 | |||
GenBank mRNA | AK035560.1 | - | 2 | 213 | ||
AK048319.1 | - | 9 | 1121 | |||
AK081984.1 | - | 2 | 139 | |||
AK139419.1 | - | 9 | 870 | |||
AK148728.1 | - | 9 | 939 | |||
AK150122.1 | - | 9 | 1091 | |||
AK150134.1 | - | 9 | 1091 | |||
AK163585.1 | - | 9 | 903 | |||
BC054533.1 | - | 9 | 1113 | 5704796 | ||
J02626.1 | - | 9 | 1058 | |||
X61434.1 | - | 1 | 28 |