Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008855.2 | NP_032881.1 | 17 | 2185 | ||
GenBank mRNA | AK051862.1 | - | 17 | 2178 | ||
BC127083.1 | - | 17 | 2004 | 40111744 | ||
X53532.1 | - | 17 | 2013 |