Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkci | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008857.3 | NP_032883.2 | 7, 8 | 865 | ||
GenBank mRNA | AK144979.1 | - | 7, 8 | 865 | ||
AK147710.1 | - | 7, 8 | 837 | |||
AK167411.1 | - | 7, 8 | 834 | |||
AK220517.1 | - | 7, 8 | 889 | |||
BC021630.1 | - | 7, 8 | 736 | 3980628 | ||
D28577.1 | - | 7, 8 | 660 |