Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013630.2 | NP_038658.2 | 15 | 6859 | ||
GenBank mRNA | U70209.1 | - | 15 | 6861 |