Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008866.2 | NP_032892.1 | 8 | 500 | ||
GenBank mRNA | AK002674.1 | - | 8 | 504 | ||
AK016021.1 | - | 604 | ||||
AK034851.1 | - | 2 | 2601 | |||
AK050549.1 | - | 8 | 477 | |||
AK146874.1 | - | 8 | 515 | |||
BC013536.1 | - | 8 | 500 | 4240573 | ||
BC052848.1 | - | 8 | 541 | 30055025 | ||
CT010201.1 | - | 8 | 479 | |||
U89352.1 | - | 8 | 479 |