Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011107.1 | NP_035237.1 | 3 | 277 | ||
GenBank mRNA | AF094610.1 | - | 3 | 270 | ||
AF097637.1 | - | 3 | 258 | |||
AF162712.1 | - | 3 | 277 | |||
AF187852.1 | - | 3 | 256 | |||
AK007730.1 | - | 3 | 271 | |||
AK007797.1 | - | 3 | 276 | |||
AK008331.1 | - | 3 | 273 | |||
AK008664.1 | - | 3 | 273 | |||
AK008668.1 | - | 3 | 273 | |||
AK008841.1 | - | 3 | 271 | |||
AK008934.1 | - | 3 | 277 | |||
AK008936.1 | - | 3 | 271 | |||
AK028104.1 | - | 3 | 274 | |||
AK028134.1 | - | 3 | 277 | |||
BC145546.1 | - | 2 | 117 | |||
BC145908.1 | - | 3 | 277 | 40131211 | ||
BC145910.1 | - | 3 | 277 | 40131213 |