Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172695.2 | NP_766283.2 | 12 | 1800 | ||
GenBank mRNA | AK019070.1 | - | 12 | 1806 | ||
AK085027.1 | - | 2 | 1247 | |||
AK166122.1 | - | 12 | 1706 | |||
BC139355.1 | - | 12 | 1799 | 8862377 | ||
BC139356.1 | - | 12 | 1799 | 8862378 | ||
BC139773.1 | - | 12 | 1710 | 40111789 | ||
DV646766.1 | - | 4 | 286 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK164744.1 | - | 2338 | Chr. - - [-] on Build |