Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008872.2 | NP_032898.2 | 14 | 1659 | ||
GenBank mRNA | AK135857.1 | - | 14 | 1650 | ||
AK135965.1 | - | 14 | 1651 | |||
AK141665.1 | - | 14 | 1648 | |||
AK159620.1 | - | 14 | 1651 | |||
AK166639.1 | - | 14 | 1649 | |||
BC011256.1 | - | 14 | 1656 | 4038404 | ||
BC057967.1 | - | 14 | 1639 | 4979664 | ||
BC061508.1 | - | 14 | 1640 | 5352018 | ||
J03520.1 | - | 14 | 1642 |